Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREML4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564198 Supplier Invitrogen™ Supplier No. PA564198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TCCFHLCCCC SWPQGAVPEE LHKHPGQTLL LQCQYSPKRG PYQPKSWCQQ TSPSRCTL Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 47%.

Positively regulates Toll-like receptor TLR7 signaling in macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREML4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TREML4
Gene Accession No. Q6UXN2
Gene Alias 5031403H21Rik; BB137214; IDCP1; IG-like domain containing protein IDCP1c; TLT4; TLT-4; TREM; TREM like transcript 4; Treml3; Treml4; trem-like transcript 4 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 4; triggering receptor expressed on myeloid cells-like 3; triggering receptor expressed on myeloid cells-like 3b; triggering receptor expressed on myeloid cells-like 4; triggering receptor expressed on myeloid cells-like protein 3; Triggering receptor expressed on myeloid cells-like protein 4; UNQ9425/PRO34675
Gene Symbols TREML4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TREML4. Recombinant protein control fragment (Product #RP-103583).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.