Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TREML4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571018
Description
This target displays homology in the following species: Human: 100%.
Positively regulates Toll-like receptor TLR7 signaling in macrophages.
Specifications
TREML4 | |
Polyclonal | |
Unconjugated | |
TREML4 | |
5031403H21Rik; BB137214; IDCP1; IG-like domain containing protein IDCP1c; TLT4; TLT-4; TREM; TREM like transcript 4; Treml3; Treml4; trem-like transcript 4 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 4; triggering receptor expressed on myeloid cells-like 3; triggering receptor expressed on myeloid cells-like 3b; triggering receptor expressed on myeloid cells-like 4; triggering receptor expressed on myeloid cells-like protein 3; Triggering receptor expressed on myeloid cells-like protein 4; UNQ9425/PRO34675 | |
Rabbit | |
Affinity chromatography | |
RUO | |
285852 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UXN2 | |
TREML4 | |
synthetic peptide directed towards the C-terminal region of human TREML4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction