Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Treml4 Rat anti-Mouse, PE, Clone: 16E5 (also known as 16E5), BD Phrmingen™
Rat Monoclonal Antibody
$268.00
Specifications
Antigen | Treml4 |
---|---|
Clone | 16E5 (also known as 16E5) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Treml4 Monoclonal antibody specifically detects Treml4 in Mouse samples. It is validated for Flow cytometrySpecifications
Treml4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
224840 | |
IgG1 κ | |
Affinity Chromatography |
16E5 (also known as 16E5) | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
Treml4; TLT-4; TLT4; IDCP1 | |
Recombinant Mouse TREML4 Extracellular Domain-Human IgG1 Fc | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title