Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREML4/TLT-4 Antibody (817914), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5500F
Description
TREML4/TLT-4 Monoclonal antibody specifically detects TREML4/TLT-4 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
TREML4/TLT-4 | |
Monoclonal | |
FITC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
285852 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
817914 | |
Flow Cytometry, CyTOF-ready | |
TLT4 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREML4/TLT-4, Gly25-Pro195, Accession # Q3LRV9 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction