Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Treponema pallidum Monoclonal Antibody (2124)
GREENER_CHOICE

Catalog No. PIMA533297
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533297 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533297 Supplier Invitrogen™ Supplier No. MA533297
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Does not cross react with Borrelia burgdorferi. Does not stain the entire bacterial cell.

This antibody reacts with ultrasonicated lysates of T. pallidum and T. reiter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Treponema pallidum
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2124
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Syphillis; t pallidum; T. pallidum
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.