Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Treponema pallidum Monoclonal Antibody (J010J)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17382
Description
The MA1-7382 antibody reacts with Treponema pallidum in bacterial and infected tissue samples. MA1-7382 has been successfully used in ELISA, Western blot and Immunofluorescence applications. The MA1-7382 antibody was raised against a Treponema pallidum preparation.
This antibody reacts with ultrasonicated lysates of T. pallidum and T. reiter.
Specifications
Treponema pallidum | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2b |
ELISA, Western Blot, Immunocytochemistry | |
J010J | |
Unconjugated | |
Syphillis; t pallidum; T. pallidum | |
Treponema pallidum preparation. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction