Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Treponema pallidum p47 Monoclonal Antibody (J25B)
GREENER_CHOICE

Catalog No. PIMA17385
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17385 Supplier Invitrogen™ Supplier No. MA17385
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7385 antibody reacts with Treponema pallidum p47 in bacterial and infected tissue samples. MA1-7385 has been successfully used in ELISA, Western blot and Immunofluorescence applications. The MA1-7385 antibody was raised against T. pallidum p47 protein.

Treponema pallidum is a gram-negative spirochaete bacterium and is considered to be metabolically crippled. There are at least four known subspecies: T. pallidum pallidum, T. pallidum pertenue, T. pallidum carateum and T. pallidum endemicum. The helical structure of T. pallidum pallidum allows it to move in a corkscrew motion through viscous mediums such as mucus. Treponema pallidum subsp. pallidum has one of the smallest bacterial genomes at 1.14 million base pairs (Mb) and has limited metabolic capabilities, reflecting its adaptation through genome reduction to the rich environment of mammalian tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Treponema pallidum p47
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone J25B
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Syphillis; t pallidum; T. pallidum
Host Species Mouse
Immunogen p47 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.