Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Treponema pallidum Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA17257
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA17257 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA17257 Supplier Invitrogen™ Supplier No. PA17257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-7257 detects Treponema pallidum. PA1-7257 has been successfully used in immunohistochemistry procedures. The PA1-7257 immunogen is highly purified Treponema pallidum.

This antibody reacts with ultrasonicated lysates of T. pallidum and T. reiter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Treponema pallidum
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Syphillis; t pallidum; T. pallidum
Host Species Rabbit
Immunogen Highly purified Treponema pallidum.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.