Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Treponema pallidum Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PIPA173101
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173101 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173101 Supplier Invitrogen™ Supplier No. PA173101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73101 antibody reacts with Treponema pallidum. PA1-73101 has been successfully used in Immunofluorescence applications. The PA1-73101 antibody was raised against Highly purified T. pallidum.

This antibody reacts with ultrasonicated lysates of T. pallidum and T. reiter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Treponema pallidum
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias Syphillis; t pallidum; T. pallidum
Host Species Rabbit
Immunogen Highly purified T. pallidum.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.