Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Treslin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538053
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538053 Supplier Invitrogen™ Supplier No. PA538053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is human small intestine tissue lysate. At least two isoforms of TICRR are known to exist.

Regulator of DNA replication and S/M and G2/M checkpoints. Regulates the triggering of DNA replication initiation via its interaction with TOPBP1 by participating in CDK2-mediated loading of CDC45L onto replication origins. Required for the transition from pre-replication complex (pre-RC) to pre-initiation complex (pre-IC). Required to prevent mitotic entry after treatment with ionizing radiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Treslin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TICRR
Gene Accession No. Q7Z2Z1
Gene Alias C15orf42; SLD3; TICRR; TOPBP1 interacting checkpoint and replication regulator; TOPBP1-interacting checkpoint and replication regulator; topBP1-interacting replication-stimulating protein; topBP1-interacting, replication-stimulating protein; Treslin
Gene Symbols TICRR
Host Species Rabbit
Immunogen 19 amino acid peptide near the carboxy terminus of human TICRR.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90381
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.