Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Treslin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583839 Supplier Invitrogen™ Supplier No. PA583839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSKVGKRCRK TSDPRRSIVE CQPDASATPG VGTADSPAAP TDSRDDQKGL SLSPQSPPER RGYPGPGLRS DWHASSPLLI TSDTEHVTLL SEAEHH Highest antigen sequence indentity to the following orthologs: Mouse - 44%, Rat - 43%.

Regulator of DNA replication and S/M and G2/M checkpoints. Regulates the triggering of DNA replication initiation via its interaction with TOPBP1 by participating in CDK2-mediated loading of CDC45L onto replication origins. Required for the transition from pre-replication complex (pre-RC) to pre-initiation complex (pre-IC). Required to prevent mitotic entry after treatment with ionizing radiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Treslin
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TICRR
Gene Accession No. Q7Z2Z1
Gene Alias C15orf42; SLD3; TICRR; TOPBP1 interacting checkpoint and replication regulator; TOPBP1-interacting checkpoint and replication regulator; topBP1-interacting replication-stimulating protein; topBP1-interacting, replication-stimulating protein; Treslin
Gene Symbols TICRR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TICRR. Recombinant protein control fragment (Product #RP-106595).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90381
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.