Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREX1 Antibody (41M5F2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222394C
Description
TREX1 Monoclonal specifically detects TREX1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TREX1 | |
Monoclonal | |
DyLight 650 | |
NP_057465.1 | |
TREX1 | |
Within the range of amino acids 211-350 of human TREX1 were used as immunogen for the antibody. | |
0.1 mL | |
DNA Repair, Editing and Processing Endonucleases | |
11277 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
41M5F2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
3'-5' exonuclease TREX1, AGS1, Aicardi-Goutieres syndrome 1, CRV, DKFZp434J0310, DNase III, DRN3, EC 3.1.11.2, HERNS, three prime repair exonuclease 1,3' repair exonuclease 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction