Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREX1 Antibody (41M5F2), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | TREX1 |
---|---|
Clone | 41M5F2 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP222394SS
![]() |
Novus Biologicals
NBP222394SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP222394
![]() |
Novus Biologicals
NBP222394 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
TREX1 Monoclonal specifically detects TREX1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TREX1 | |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
DNA Repair, Editing and Processing Endonucleases | |
NP_057465.1 | |
11277 | |
Within the range of amino acids 211-350 of human TREX1 were used as immunogen for the antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
41M5F2 | |
Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
3'-5' exonuclease TREX1, AGS1, Aicardi-Goutieres syndrome 1, CRV, DKFZp434J0310, DNase III, DRN3, EC 3.1.11.2, HERNS, three prime repair exonuclease 1,3' repair exonuclease 1 | |
TREX1 | |
IgG2b κ | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title