Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520665
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520665 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520665 Supplier Invitrogen™ Supplier No. PA520665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human spleen tissue lysate. PA5-20665 can be used with blocking peptide PEP-0785.

Trex1 (ATRIP) is the major human 3' to 5' exonuclease which is required for checkpoint signaling after DNA damage. It is ubiquitously expressed, binds to single stranded DNA coated with replication protein A that accumulates at sites of DNA damage and recruits the ataxia telangiectasia and Rad3 related protein (ATR), a checkpoint kinase, to sites of DNA damage and replication stress. Trex1 is required for ATR expression. This gene uses two different open reading frames. The upstream ORF encodes proteins which interact with ATR and localize to intranuclear foci induced by DNA damage and are essential components of the DNA damage checkpoint. The downstream ORF encodes proteins with 3' to 5' exonuclease activity and may be a subunit of human DNA polymerase III. Multiple transcript variants encoding different isoforms have been found. Mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREX1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TREX1
Gene Accession No. Q91XB0, Q9NSU2
Gene Alias 1661; 3' repair exonuclease 1; 3'-5' exonuclease TREX1; AGS1; AU041952; CRV; Deoxyribonuclease III; DNase III; DRN3; HERNS; RGD1309596; three prime exonuclease 1; three prime repair exonuclease 1; three-prime repair exonuclease 1; Trex1; trophoblast expressed 1
Gene Symbols TREX1
Host Species Rabbit
Immunogen A 15 amino acid peptide near the center of human TREX1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049583, 11277, 22040
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.