Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111063
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111063 100 μL
PIPA5111242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111063 Supplier Invitrogen™ Supplier No. PA5111063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDAHARPFGT IRPMYGVTAS ARTKPRPSAV TTTAHLATTR NTSPSLGESR GTKDLPPVKD PGALSRE Highest antigen sequence indentity to the following orthologs: Rat - 54%, Mouse - 54%.

Trex1 (ATRIP) is the major human 3' to 5' exonuclease which is required for checkpoint signaling after DNA damage. It is ubiquitously expressed, binds to single stranded DNA coated with replication protein A that accumulates at sites of DNA damage and recruits the ataxia telangiectasia and Rad3 related protein (ATR), a checkpoint kinase, to sites of DNA damage and replication stress. Trex1 is required for ATR expression. This gene uses two different open reading frames. The upstream ORF encodes proteins which interact with ATR and localize to intranuclear foci induced by DNA damage and are essential components of the DNA damage checkpoint. The downstream ORF encodes proteins with 3' to 5' exonuclease activity and may be a subunit of human DNA polymerase III. Multiple transcript variants encoding different isoforms have been found. Mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREX1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TREX1
Gene Accession No. Q9NSU2
Gene Alias 1661; 3' repair exonuclease 1; 3'-5' exonuclease TREX1; AGS1; AU041952; CRV; Deoxyribonuclease III; DNase III; DRN3; HERNS; RGD1309596; three prime exonuclease 1; three prime repair exonuclease 1; three-prime repair exonuclease 1; Trex1; trophoblast expressed 1
Gene Symbols TREX1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TREX1. Recombinant protein control fragment (Product #RP-96493).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.