Learn More
Invitrogen™ TREX1 Polyclonal Antibody

Description
Immunogen sequence: VDAHARPFGT IRPMYGVTAS ARTKPRPSAV TTTAHLATTR NTSPSLGESR GTKDLPPVKD PGALSRE Highest antigen sequence indentity to the following orthologs: Rat - 54%, Mouse - 54%.
Specifications
Specifications
| Antigen | TREX1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TREX1 |
| Gene Accession No. | Q9NSU2 |
| Gene Alias | 1661; 3' repair exonuclease 1; 3'-5' exonuclease TREX1; AGS1; AU041952; CRV; Deoxyribonuclease III; DNase III; DRN3; HERNS; RGD1309596; three prime exonuclease 1; three prime repair exonuclease 1; three-prime repair exonuclease 1; Trex1; trophoblast expressed 1 |
| Gene Symbols | TREX1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.