Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TREX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562906 Supplier Invitrogen™ Supplier No. PA562906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RAETFVFLDL EATGLPSVEP EIAELSLFAV HRSSLENPEH DESGALVLP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

TREX2 is one of two major human 3' to 5' exonucleases which are required for checkpoint signaling after DNA damage. While the related protein TREX1 is required for ATR expression and mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis, less is known about TREX2. Like TREX1, TREX2 is ubiquitously expressed in all tissues examined. X-ray crystallography studies of TREX2-single strand DNA complexes revealed that TREX2 binds DNA as a dimer and may act to displace the second DNA strand, suggesting that TREX2 unzips and denatures double stranded DNA and feed the substrate DNA strand into the TREX2 active site. At least two isoforms of TREX2 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TREX2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TREX2
Gene Accession No. Q9BQ50
Gene Alias 3'-5' exonuclease TREX2; 3'-5' exonuclease TREX2 long form; RGD1562245; Three prime repair exonuclease 2; Trex2
Gene Symbols TREX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TREX2. Recombinant protein control fragment (Product #RP-101552).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11219
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.