Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF-1 Antibody (57-6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280993
Description
TRF-1 Monoclonal specifically detects TRF-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, ICC/IF (Negative).Specifications
TRF-1 | |
Monoclonal | |
1.5 mg/mL | |
Western Blot : 0.5 μg/mL, Simple Western : 1:200, Flow Cytometry : 1 ug per million cells, ELISA 1:100 - 1:2000, ICC/IF (Negative) | |
NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
57-6 | |
Unconjugated | |
Tris-Glycine, 0.15M NaCl | |
TERF1 | |
Recombinant human TRF1 [UniProt# P54274] | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
7013 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction