Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-1 Antibody (57-6), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$207.00 - $499.50

Specifications

Antigen TRF-1
Clone 57-6
Concentration 1.0 mg/ml
Dilution Western Blot 0.5 ug/ml, Simple Western 1:200, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, ICC/IF (Negative)
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11068281T
SDP
View Documents
Novus Biologicals
NB11068281SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NB11068281
SDP
View Documents
Novus Biologicals
NB11068281
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

TRF-1 Monoclonal specifically detects TRF-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, ICC/IF (Negative).
Specifications

Specifications

TRF-1
1.0 mg/ml
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Unconjugated
Mouse
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
P54274
7013
Recombinant human TRF1 [UniProt# P54274]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
57-6
Western Blot 0.5 ug/ml, Simple Western 1:200, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, ICC/IF (Negative)
Monoclonal
Purified
RUO
Human, Mouse, Rat, Monkey
NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1
TERF1
IgG1 κ
Protein G purified
60 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.