Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF-1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100891
Description
TRF-1 Polyclonal specifically detects TRF-1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
| TRF-1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_059523.2) and isoform 2 (NP_003209.2). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_003209 | |
| TERF1 | |
| Peptide with sequence C-TMKKLKLISSDSED corresponding to C-Terminus according to NP_003209.2, NP_059523.2. | |
| 0.1 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| 7013 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction