Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF-2 Antibody (4A794.15), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056506MFV610
Description
TRF-2 Monoclonal antibody specifically detects TRF-2 in Human, Mouse, Rat, Deer, Marsupial samples. It is validated for Flow CytometrySpecifications
TRF-2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Deer, Marsupial | |
Purified |
Flow Cytometry | |
4A794.15 | |
Flow Cytometry | |
Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF2, TRBF2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 | |
This TRF-2 Antibody (4A794.15) was developed against Baculovirus expressed whole length TRF2 protein, used for immunizing mice and splenocytes used to generate the hybridoma clone (NP_005643). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators | |
7014 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction