Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF-2 Antibody (SR2115), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32190550UL
Description
TRF-2 Monoclonal antibody specifically detects TRF-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
TRF-2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR2115 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF2, TRBF2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 | |
A synthesized peptide derived from human TRF-2 (Uniprot #: Q15554) | |
50 μg | |
Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators | |
7014 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction