Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRF1 Monoclonal Antibody (4E4)

Catalog No. PIMA531596
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531596 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531596 Supplier Invitrogen™ Supplier No. MA531596

Mouse Monoclonal Antibody

Keep as concentrated solution.

This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRF1
Applications ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4E4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with no preservative
Gene TERF1
Gene Accession No. P54274
Gene Alias FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1
Gene Symbols TERF1
Host Species Mouse
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7013
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.