Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRF1 Monoclonal Antibody (57-6)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA146375

Catalog No. PIMA146375


Only null left
Add to Cart

Description

Description

This antibody will not work in ICC/IF applications. Suggested positive control: HeLa nuclear extracts, antigen standard for TERF1 (transient overexpression lysate), Hela whole cell extract.

This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRF1
Monoclonal
1 mg/mL
TBS with 0.05% sodium azide
P54274, P70371
TERF1
Recombinant TRF-1.
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG1 κ
ELISA, Flow Cytometry, Western Blot, Western Blot
57-6
Unconjugated
TERF1
FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1
Mouse
Protein G
RUO
21749, 297758, 7013
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.