Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587358 Supplier Invitrogen™ Supplier No. PA587358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse eye; Cellular Location: Chromosome, Cytoplasm, Nucleus, cytoskeleton, spindle, telomere Immunogen sequence: VGAPEEEEEE EEDAGLVAEA EAVAAGWMLD FLCLSLCRAF RDGRSEDFRR TRNSAEAIIH GLSSLTACQL RTIYICQFLT RIAAGKTLDA QFENDERITP LESALMIWGS IEKEHDKLHE EIQNLIKIQA IAVCMENGNF KEAEEVFERI FGDPNSHMPF KSKLLMIISQ KDTFHSFFQH FSYNHMMEKI KSYVNYVLSE KSSTFLMKAA AKVVESKRTR TITSQ.

This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TERF1
Gene Accession No. P54274
Gene Alias FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1
Gene Symbols TERF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 52-275 of human TERF1 (NP_0595232?).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297758, 7013
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.