Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRF1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142525
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoform 1 (NP_059523.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
Specifications
TRF1 | |
Polyclonal | |
Unconjugated | |
TERF1 | |
FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
7013 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P54274 | |
TERF1 | |
Peptide with sequence C-NLDTRKSVSDKQS. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction