Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF2 Mouse anti-Human, Clone: T2JKM3, MilliporeSigma™

Catalog No. MABE1135100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1135-100 100 μg
MABE1135-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1135-100 Supplier MilliporeSigma Supplier No. MABE1135100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TRF2, clone T2JKM3 is a highly specific mouse monoclonal antibody that targets Telomeric repeat-binding factor 2 (TRF2) and has been tested for use in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRF2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone T2JKM3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TERF2,TRF2,TRBF2
Host Species Mouse
Immunogen Tagged full length recombinant human Telomeric repeat-binding factor 2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005643
Test Specificity Clone T2JKM3 detects Telomeric repeat-binding factor 2 in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.