Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRF2 Recombinant Rabbit Monoclonal Antibody (JE37-57)
GREENER_CHOICE

Catalog No. FEMA550791
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550791 Supplier Invitrogen™ Supplier No. MA550791
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: SiHa cell lysate, Jurkat cell lysate, human colon carcinoma tissue, human testis tissue, SiHa. Predicted band size: 60 kDa Subcellular Location: Nucleus, Chromosome, telomere.

TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length. TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRF2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE37-57
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TERF2
Gene Accession No. Q15554
Gene Alias Telomeric DNA-binding protein; telomeric repeat binding factor 2; telomeric repeat binding protein 2; telomeric repeat-binding factor 2; TERF2; TRBF2; TRF2; TTAGGG repeat-binding factor 2
Gene Symbols TERF2
Host Species Rabbit
Immunogen Recombinant protein within Human TRF2 aa 1-100/542.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7014
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.