Learn More
TRGC2 Polyclonal Antibody

Description
TRGC2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | TRGC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TRGC2 |
| Gene Accession No. | P03986 |
| Gene Alias | T cell receptor gamma constant 2; TCRGC2; TRGC2; TRGC2(2X); TRGC2(3X) |
| Gene Symbols | TRGC2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.