Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589851 Supplier Invitrogen™ Supplier No. PA589851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, BXPC-3, A-549, DU145, Mouse brain, Rat testis Immunogen sequence: WLAYAVPKRQ HPGRRLADPK AQRSWEEEEE EEEREEDLMP EKRQHPGKRA LGGPCGPQGA YGQAGLLLGL LDDLSRSQGA EEKRQHPGRR AAWVREPLEE.

This gene encodes a member of the thyrotropin-releasing hormone family. Cleavage of the encoded proprotein releases mature thyrotropin-releasing hormone, which is a tripeptide hypothalamic regulatory hormone. The human proprotein contains six thyrotropin-releasing hormone tripeptides. Thyrotropin-releasing hormone is involved in the regulation and release of thyroid-stimulating hormone, as well as prolactin. Deficiency of this hormone has been associated with hypothalamic hypothyroidism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRH
Gene Accession No. P01150, P20396, Q62361
Gene Alias Prothyroliberin; Pro-thyrotropin-releasing hormone; protirelin; Pro-TRH; THR; Thyroliberin; thyrotropin releasing hormone; thyrotropin-releasing factor; Thyrotropin-releasing hormone; TRF; Trh; TRH01; TSH-releasing factor
Gene Symbols TRH
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 143-242 of human TRH (NP_0090481).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22044, 25569, 7200
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.