Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRHDE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547429
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547429 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547429 Supplier Invitrogen™ Supplier No. PA547429
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a member of the peptidase M1 family. TRHDE is an extracellular peptidase that specifically cleaves and inactivates the neuropeptide thyrotropin-releasing hormone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRHDE
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TRHDE
Gene Accession No. Q10836, Q8K093
Gene Alias 9330155P21Rik; PAP-II; PGPEP2; pyroglutamyl aminopeptidase II; pyroglutamyl-peptidase II; Thyroliberinase; thyrotropin releasing hormone degrading enzyme; thyrotropin-releasing hormone degrading ectoenzyme; thyrotropin-releasing hormone degrading enzyme; thyrotropin-releasing hormone-degrading ectoenzyme; Trhde; TRH-DE; TRH-degrading ectoenzyme; TRH-degrading enzyme; TRH-specific aminopeptidase; UNQ2507/PRO5995
Gene Symbols TRHDE
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TRH-degrading Ectoenzyme/TRHDE Arg64-His1025.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237553, 366894
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.