Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRHDE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113501 Supplier Invitrogen™ Supplier No. PA5113501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRHDE Polyclonal Antibody for Western Blot, ELISA

This gene encodes a member of the peptidase M1 family. TRHDE is an extracellular peptidase that specifically cleaves and inactivates the neuropeptide thyrotropin-releasing hormone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRHDE
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.01M PBS with 50% glycerol and no preservative; pH 7.4
Gene TRHDE
Gene Accession No. Q8K093, Q9UKU6
Gene Alias 9330155P21Rik; PAP-II; PGPEP2; pyroglutamyl aminopeptidase II; pyroglutamyl-peptidase II; Thyroliberinase; thyrotropin releasing hormone degrading enzyme; thyrotropin-releasing hormone degrading ectoenzyme; thyrotropin-releasing hormone degrading enzyme; thyrotropin-releasing hormone-degrading ectoenzyme; Trhde; TRH-DE; TRH-degrading ectoenzyme; TRH-degrading enzyme; TRH-specific aminopeptidase; UNQ2507/PRO5995
Gene Symbols TRHDE
Host Species Rabbit
Immunogen Recombinant Human Thyrotropin-releasing hormone-degrading ectoenzyme protein (62-200AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237553, 29953
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.