Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRHR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534278
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Elephant, Horse (100%).
TRH-R is a 398 amino acid integral membrane protein with six transmembrane domains and belongs to G-protein coupled receptor 1 family. It acts as a receptor for the signaling hormone TRH and activates the inositol phospholipid-calcium-protein kinase C transduction pathway upon binding with TRH. Reports suggest that TRH-R is a neurotransmitter and neuro regulator that controls dimerization of the TRH receptor and potentiates hormone-induced receptor trafficking. It is known to widely express in the thyrotroph cells of the anterior pituitary.
Specifications
| TRHR | |
| Polyclonal | |
| Unconjugated | |
| TRHR | |
| MGC141920; Thyroliberin receptor; thyrotropin releasing hormone receptor; thyrotropin-releasing hormone receptor; TRH receptor; TRH1 Receptor; Trhr; TRH-R; TRH-R1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100056618, 22045, 25570, 7201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P21761, P34981, Q01717 | |
| TRHR | |
| Synthetic 16 amino acid peptide from C-terminus of human TRH Receptor. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction