Learn More
Description
Application: Flow cytometry, fluorescence microscopy.
Properties and Photophysical Data: Suitable for multiplexing and can be combined with DAPI (Cat. No. 5748) counter-staining. Excitation and emission maxima (λ) are 663 and 684 nm, respectively.
Specifications
Specifications
| Quantity | 100 μg |
| Target | Fluorescent Receptor Probes |
| Molecular Formula | C103H162N16O44S4 |
| Purity | 0.9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

