Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tri-Methyl Lysine Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116919
Description
Positive Samples: HeLa, MCF7, NIH/3T3, C6.
Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.
Specifications
| Tri-Methyl Lysine | |
| Polyclonal | |
| Unconjugated | |
| Trimethyllysine | |
| A synthetic peptide corresponding to a sequence containing Tri-methylated K. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ChIP Assay, ELISA, Immunoprecipitation, Western Blot | |
| 1.04 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Chemical, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction