Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tri-n-butylphenyltin, 97%

Manufacturer:  Alfa Aesar™ H5559603

Quantity 1g

 View more versions of this product

Catalog No. AAH5559603


  Check Availability
Add to cart

Specifications

Description

Applications
It is applied in chemical research.

Solubility
Sparingly Soluble (9.9E-5 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
Description & Specifications

Chemical Identifiers

960-16-7
367.14
UN2788
Phenyltri-n-butyltin; Tri-n-butylphenylstannane
C6H5Sn[(CH2)3CH3]3
MFCD00134394
3610571
Tri-n-butylphenyltin

Specifications

125° to 128°C (0.14mmHg)
1g
Colorless
Liquid
367.14
UN2788
Phenyltri-n-butyltin; Tri-n-butylphenylstannane
97%
960-16-7
1.125g/mL
1.5160
C6H5Sn[(CH2)3CH3]3
MFCD00134394
3610571
Tri-n-butylphenyltin

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit