Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin chloride, 96%

Catalog No. p-7080977
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 1461-22-9 | C12H27ClSn | 325.508 g/mol

Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.

Solubility
Miscible with alcohol, heptane, benzene and toluene. Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-22-9
Molecular Formula C12H27ClSn
Molecular Weight (g/mol) 325.508
MDL Number MFCD00000521
InChI Key GCTFWCDSFPMHHS-UHFFFAOYSA-M
Synonym tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride
PubChem CID 15096
ChEBI CHEBI:79734
IUPAC Name tributyl(chloro)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Cl

Specifications

Melting Point -19°C
Density 1.2
Boiling Point 171°C to 173°C (25 mmHg)
Flash Point 120°C (248°F)
Refractive Index 1.491
Quantity 50 g
UN Number UN2788
Beilstein 3535715
Solubility Information Miscible with alcohol,heptane,benzene and toluene. Immiscible with water.
Formula Weight 325.51
Percent Purity 96%
Chemical Name or Material Tri-n-butyltin chloride
Show More Show Less
Hazard Category H301-H312-H315-H319-H360FD-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 215-958-7
RTECSNumber WH6820000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.