Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin methoxide, 97%

Catalog No. p-7021140
Click to view available options
Quantity:
100 g
250 g
1000 g

Catalog No. AAA1335522

Supplier: Thermo Scientific Chemicals A1335522

Only null left
Add to Cart

CAS: 1067-52-3 | C13H30OSn | 321.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-52-3
Molecular Formula C13H30OSn
Molecular Weight (g/mol) 321.092
MDL Number MFCD00009419
InChI Key KJGLZJQPMKQFIK-UHFFFAOYSA-N
Synonym tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl
PubChem CID 16683411
IUPAC Name tributyl(methoxy)stannane
SMILES CCCC[Sn](CCCC)(CCCC)OC

Specifications

Density 1.12
Boiling Point 124°C to 126°C (4 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.474
Quantity 100 g
UN Number UN2788
Beilstein 3600960
Sensitivity Moisture sensitive
Formula Weight 321.09
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin methoxide
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 213-933-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.