Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin trifluoromethanesulfonate, tech. 90%, Thermo Scientific™

Catalog No. AAH5519203
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68725-14-4
MDL Number MFCD00043324
Synonym Tri-n-butyltin triflate

Specifications

Color Brown to White
Flash Point 113°C (235°F)
Linear Formula CF3SO3Sn[(CH2)3CH3]3
Quantity 1 g
UN Number UN2923
Beilstein 5818090
Sensitivity Moisture sensitive
Formula Weight 439.1
Percent Purity 90%
Grade Technical
Physical Form Solid
Chemical Name or Material Tri-n-butyltin trifluoromethanesulfonate
Show More Show Less
Hazard Category H301-H312-H314-H335-H372
Hazard Statement GHS H Statement
H301-H372-H314-H318-H312
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.