Learn More
Tri-n-hexylamine, 96%
Description
Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, acids, acid chloridres, acid anhydrides and chloroformates.

Specifications
Specifications
Melting Point | <-75°C |
Density | 0.798 |
Boiling Point | 263°C to 265°C |
Flash Point | 144°C (291°F) |
Refractive Index | 1.4415 |
Quantity | 250 g |
UN Number | UN3082 |
Beilstein | 1755828 |
Solubility Information | Slightly miscible with water. |
Formula Weight | 269.52 |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
EINECSNumber | 203-062-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.