Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-octylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
1116-76-3
C24H51N
353.66
XTAZYLNFDRKIHJ-UHFFFAOYSA-N
trioctylamine,tri-n-octylamine,tricaprylamine,tricaprylylamine,1-octanamine, n,n-dioctyl,tri-n-caprylylamine,alamine 336,alamine 336s,alamine 3365,farmin 08
14227
N,N-dioctyloctan-1-amine
CCCCCCCCN(CCCCCCCC)CCCCCCCC
1116-76-3
C24H51N
353.66
XTAZYLNFDRKIHJ-UHFFFAOYSA-N
trioctylamine,tri-n-octylamine,tricaprylamine,tricaprylylamine,1-octanamine, n,n-dioctyl,tri-n-caprylylamine,alamine 336,alamine 336s,alamine 3365,farmin 08
14227
N,N-dioctyloctan-1-amine
CCCCCCCCN(CCCCCCCC)CCCCCCCC
Boiling Point 365.0°C to 367.0°C
Color White
Quantity 500mL
Flash Point 163°C
Melting Point -34.0°C
Refractive Index 1.4480 to 1.4500
Water 0.1% max.
Packaging Glass bottle
Specific Gravity 0.809
Infrared Spectrum Authentic
Density 0.8090g/mL
Assay Percent Range 97%
Linear Formula [CH3(CH2)7]3N
Beilstein 04,196
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 353.66
Physical Form Low Melting Crystalline Mass
Percent Purity 97%
Chemical Name or Material Tri-n-octylamine
Show More Show Less