Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-octylphosphine 85.0+%, TCI America™
SDP

Catalog No. T0503100ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL

Chemical Identifiers

CAS 4731-53-7
Molecular Formula C24H51P
Molecular Weight (g/mol) 370.646
MDL Number MFCD00015298
InChI Key RMZAYIKUYWXQPB-UHFFFAOYSA-N
Synonym trioctylphosphine, tri-n-octylphosphine, phosphine, trioctyl, tri-n-octyl phosphine, tri-octylphosphine, trioctyl phosphine, tri degrees ctylphosphine, acmc-209k8q
PubChem CID 20851
IUPAC Name trioctylphosphane
SMILES CCCCCCCCP(CCCCCCCC)CCCCCCCC

Specifications

Color Yellow
Boiling Point 234°C
Quantity 100 mL
UN Number 1760
Formula Weight 370.65
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Tri-n-octylphosphine
EINECSNumber (2)-3735
RTECSNumber SZ3450000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.