Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tri-p-cresyl Phosphate 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
| Melting Point | 77°C |
| Color | White-Yellow |
| Boiling Point | 430°C |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 368.37 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tri-p-cresyl Phosphate |
Safety and Handling
| EINECSNumber | (3)-2522&(3)-2613&(3)-3363 |
| RTECSNumber | TC9330000 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction