Learn More
Tri-tert-butylphosphine, 98%
Description
Tri-tert-butylphosphine is used primarily in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations and other asymmetric reactions as well as transition metal coupling reactions. This reagent is also used in the preparation of other phosphines in catalytic polymerization reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 30°C |
| Color | White |
| Boiling Point | 102°C to 103°C (13 mmHg) |
| Flash Point | -16°C (-3°F) |
| Odor | Pungent |
| Linear Formula | [(CH3)3C]3P |
| Quantity | 1 g |
| UN Number | UN2846 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Insoluble in water. Soluble in alcohol and organic solvents |
| Show More |
Safety and Handling
| Hazard Category | H228-H250-H314-H335 |
| Hazard Statement | GHS H Statement H250-H252-H314-H318 Catches fire spontaneously if exposed to air. Self-heating in large quantities may catch fire. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P222-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 237-266-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.