Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylene glycol monomethyl ether

Catalog No. 3172922L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-729-22L 2 L
31-729-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-729-22L Supplier Sigma Aldrich Supplier No. 3172922L
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylene glycol monomethyl ether can be used as a reagent and solvent for applications such as: modification of anthraquinone material for redox flow batteriespreparation of polymeric electrolyte for electrochemical devices,formation of the binary system of polyethylene glycol for absorption of silica.

Specifications

CAS 112-35-6
Density 1.026 g/mL (at 25°C)
Boiling Point 122°C
Molecular Formula C7H16O4
Linear Formula CH3(OCH2CH2)3 OH
Refractive Index n20/D 1.439 (literature)
Quantity 2 L
MDL Number MFCD00002875
Synonym Methyltriglycol
Molecular Weight (g/mol) 164.2
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.