Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacetin, FCC, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18607500
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle

T1200, 102-76-1, C9H14O6

Spectrum™ Chemical Triacetin, FCC is widely used as humectant, emlusifier, binder in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 102-76-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
InChI Key URAYPUMNDPQOKB-UHFFFAOYSA-N
IUPAC Name 1,3-bis(acetyloxy)propan-2-yl acetate
SMILES CC(=O)OCC(COC(C)=O)OC(C)=O

Specifications

Acidity To pass test
CAS Max % 1
Boiling Point 258°C
Packaging Amber Glass Bottle
Refractive Index 1.429-1.431
Quantity 500 mL
Percent Purity 98.5%
Grade FCC
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.