Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacontane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015022 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 mg
1 g

Catalog No. 11-101-5022

Supplier: MilliporeSigma Supelco 902701G

Only null left
Add to Cart
Add to Cart

Triacontane is an acyclic linear alkane that physically appears like a paraffin like solid, which occurs like a waxy coat on tobacco leaves.

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 638-68-6
Physical Form Neat
Linear Formula CH3(CH2)28 CH3
MDL Number MFCD00009410
Melting Point 64°C to 67°C; 64°C to 67°C
Boiling Point 258°C to 259°C (3 mmHg)
Type Analytical Standard
Quantity 1 g
Formula Weight 422.81
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.