Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacontanoic Acid Methyl Ester, 98%, MP Biomedicals™

Catalog No. ICN15695980 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 629-83-4
Molecular Formula C31H62O2
Molecular Weight (g/mol) 466.84
InChI Key BIRUBGLRQLAEFF-UHFFFAOYSA-N
Synonym methyl triacontanate, methyl melissate, melissic acid methyl ester, methyl melissicate, triacontanoic acid methyl ester, triacontanoic acid, methyl ester, n-triacontanoic acid methyl ester, triacontanoic acid methylester, methyltriacontanate, acmc-1b46i
PubChem CID 12400
IUPAC Name methyl triacontanoate
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Color White
Assay Percent Range 98%
Quantity 100 mg
Formula Weight 466.8
Physical Form Powder
Chemical Name or Material Triacontanoic Acid Methyl Ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.