Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Triacsin C
GREENER_CHOICESDP

Catalog No. 2472100U Shop All R&D Systems Products
Change view
Click to view available options
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
24-721-00U 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-721-00U Supplier Tocris Bioscience™ Supplier No. 2472/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of acyl-CoA synthetase

Triacsin C is an inhibitor of acyl-CoA synthetase. Inhibits lipid synthesis and cell proliferation in Raji lymphoma cells. Induces apoptosis in cancer cell lines and inhibits human lung cancer cell xenograft tumor growth in mice.

Specifications

Quantity 100 μg
Product Type Triacsin C
Purity >95%
Molecular Weight (g/mol) 207.27
Inhibitors Triacsin C

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.