Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triad1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501563755
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3755 Supplier Bethyl Laboratories, Inc Supplier No. A302896A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

E3 ubiquitin-protein ligase mediating ′Lys-48′-and ′Lys-63′-linked polyubiquitination and subsequent proteasomal degradation of modified proteins. May play a role in myelopoiesis.

Specifications

Antigen Triad1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ARIH2
Gene Accession No. O95376, Q9Z1K6
Gene Alias ARIH2; ariadne homolog 2 (Drosophila); ARI2; TRIAD1; all-trans retinoic acid inducible RING finger; ariadne-2 (D. melanogaster) homolog (all-trans retinoic acid inducible RING finger); FLJ10938; FLJ33921; OTTHUMP00000210388; ariadne homolog 2
Gene Symbols Arih2
Host Species Rabbit
Immunogen Between 453 and 493
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10425, 23807
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.